Product Details

SNP ID
rs2782225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:119759450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCAACCTGAACAACAACAGCAG[C/T]GGCACCACTGCGTGGAGGGCCGCGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SOWAHD PubMed Links

Gene Details

Gene
SOWAHD
Gene Name
sosondowah ankyrin repeat domain family member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105576.2 838 Silent Mutation AGC,AGT S261S NP_001099046.1

View Full Product Details