Product Details

SNP ID
rs2919360
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65078015 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCTCTTCACGGCCGTGGTGCGCGG[C/T]GACACCGTCTATACGGTCAACCGCA
Phenotype
MIM: 613727
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KBTBD13 PubMed Links

Gene Details

Gene
KBTBD13
Gene Name
kelch repeat and BTB domain containing 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101362.2 1200 Silent Mutation GGC,GGT G400G NP_001094832.1
Gene
RASL12
Gene Name
RAS like family 12
There are no transcripts associated with this gene.

View Full Product Details