Product Details

SNP ID
rs2904015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56371260 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTGGCAGGCCATGTCTCACCAAA[C/T]GCAGGTGTCCATAACAGCTGTTTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF542P PubMed Links

Gene Details

Gene
ZNF542P
Gene Name
zinc finger protein 542, pseudogene
There are no transcripts associated with this gene.

Gene
ZSCAN5A
Gene Name
zinc finger and SCAN domain containing 5A
There are no transcripts associated with this gene.

View Full Product Details