Product Details

SNP ID
rs2925141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18111799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCCCAGGCCAGCCACTGTGCCC[A/G]GCTCTCCGCTATCTCATGTAATGCC
Phenotype
MIM: 602986 MIM: 602666
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DRG2 PubMed Links

Gene Details

Gene
DRG2
Gene Name
developmentally regulated GTP binding protein 2
There are no transcripts associated with this gene.

Gene
MYO15A
Gene Name
myosin XVA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016239.3 Intron NP_057323.3
XM_011523918.2 Intron XP_011522220.1
XM_017024714.1 Intron XP_016880203.1
XM_017024715.1 Intron XP_016880204.1

View Full Product Details