Product Details

SNP ID
rs2232795
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3021001 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGAGTCACCCGCCGGCGGCAGGGG[A/G]GTTGTGGGGACCTGACCCCCCCCAC
Phenotype
MIM: 617003 MIM: 615798 MIM: 615799 MIM: 615403 MIM: 605914
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BICDL2 PubMed Links

Gene Details

Gene
BICDL2
Gene Name
BICD family like cargo adaptor 2
There are no transcripts associated with this gene.

Gene
CLDN6
Gene Name
claudin 6
There are no transcripts associated with this gene.

Gene
CLDN9
Gene Name
claudin 9
There are no transcripts associated with this gene.

Gene
HCFC1R1
Gene Name
host cell factor C1 regulator 1
There are no transcripts associated with this gene.

Gene
THOC6
Gene Name
THO complex 6
There are no transcripts associated with this gene.

Gene
TNFRSF12A
Gene Name
TNF receptor superfamily member 12A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016639.2 Intron NP_057723.1

View Full Product Details