Product Details

SNP ID
rs2233979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31111918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACTCCCCAGGCAAAGGGCCACT[C/G]CCCGGAGCGAGGGCCGCAGCACTGG
Phenotype
MIM: 611401 MIM: 602593 MIM: 613525
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C6orf15 PubMed Links

Gene Details

Gene
C6orf15
Gene Name
chromosome 6 open reading frame 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014070.2 441 Silent Mutation GGC,GGG G147G NP_054789.2
Gene
CDSN
Gene Name
corneodesmosin
There are no transcripts associated with this gene.

Gene
PSORS1C1
Gene Name
psoriasis susceptibility 1 candidate 1
There are no transcripts associated with this gene.

View Full Product Details