Product Details
- SNP ID
-
rs2234013
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:141790419 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTGGCAGTGGTTGAATTTCTCATC[A/G]GTTTAATTGGAAATGGAAGCCTGGT
- Phenotype
-
MIM: 605062
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TAS2R5
PubMed Links
Gene Details
- Gene
- TAS2R5
- Gene Name
- taste 2 receptor member 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018980.2 |
203 |
Missense Mutation |
AGT,GGT |
S20G |
NP_061853.1 |
View Full Product Details