Product Details

SNP ID
rs2234151
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:2556334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCGGAGTGGACTGGAATGGTGCA[G/C]GGGGAGAACTCGCCCCTCCCATCGG
Phenotype
MIM: 602746
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC115110 PubMed Links

Gene Details

Gene
LOC115110
Gene Name
uncharacterized LOC115110
There are no transcripts associated with this gene.

Gene
TNFRSF14
Gene Name
TNF receptor superfamily member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297605.1 Intron NP_001284534.1
NM_003820.3 Intron NP_003811.2
XM_006711019.2 Intron XP_006711082.1
XM_011542383.2 Intron XP_011540685.1
XM_017002718.1 Intron XP_016858207.1
XM_017002719.1 Intron XP_016858208.1
XM_017002720.1 Intron XP_016858209.1
XM_017002721.1 Intron XP_016858210.1

View Full Product Details