Product Details

SNP ID
rs2234154
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:2556562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCACACCGAGGCGGATTCTCTT[T/C]CTCTTTCTCTTTCTCTTCTGGCCCA
Phenotype
MIM: 602746
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
LOC100996583 PubMed Links

Gene Details

Gene
LOC100996583
Gene Name
uncharacterized LOC100996583
There are no transcripts associated with this gene.

Gene
LOC115110
Gene Name
uncharacterized LOC115110
There are no transcripts associated with this gene.

Gene
TNFRSF14
Gene Name
TNF receptor superfamily member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297605.1 198 UTR 5 NP_001284534.1
NM_003820.3 198 UTR 5 NP_003811.2
XM_006711019.2 198 Intron XP_006711082.1
XM_011542383.2 198 Intron XP_011540685.1
XM_017002718.1 198 Intron XP_016858207.1
XM_017002719.1 198 Intron XP_016858208.1
XM_017002720.1 198 Intron XP_016858209.1
XM_017002721.1 198 Intron XP_016858210.1

View Full Product Details