Product Details

SNP ID
rs2953278
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:201451761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCGTTCCCAGCGGGTAGCCTGGG[A/G]CTGGTGCAGAGTTCCAAGCCCACGG
Phenotype
MIM: 607333 MIM: 607334
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STRADB PubMed Links

Gene Details

Gene
STRADB
Gene Name
STE20-related kinase adaptor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206864.1 Intron NP_001193793.1
NM_018571.5 Intron NP_061041.2
XM_005246669.1 Intron XP_005246726.1
XM_017004438.1 Intron XP_016859927.1
Gene
TRAK2
Gene Name
trafficking kinesin protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015049.2 Intron NP_055864.2
XM_011511690.1 Intron XP_011509992.1
XM_017004772.1 Intron XP_016860261.1

View Full Product Details