Product Details

SNP ID
rs2227090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:85279377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACTAATGCAGAGACACACACACA[A/C]AAAAAAAACGGCTTTGAGTTGTAAT
Phenotype
MIM: 300603 MIM: 314990
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
POF1B PubMed Links

Gene Details

Gene
POF1B
Gene Name
premature ovarian failure, 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307940.1 1873 Intron NP_001294869.1
NM_024921.3 1873 UTR 3 NP_079197.3
XM_005262203.3 1873 UTR 3 XP_005262260.1
Gene
ZNF711
Gene Name
zinc finger protein 711
There are no transcripts associated with this gene.

View Full Product Details