Product Details
- SNP ID
-
rs2236699
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:13826008 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TCTTCGTGTGCAGCCTGGCAGTGGC[G/T]GACATGCTGGTGAGCATGTCCAGTG
- Phenotype
-
MIM: 600042
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MC5R
PubMed Links
Gene Details
- Gene
- MC5R
- Gene Name
- melanocortin 5 receptor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005913.2 |
465 |
Silent Mutation |
GCG,GCT |
A81A |
NP_005904.1 |
View Full Product Details