Product Details

SNP ID
rs2227325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40016019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGACCTTGGTGGGGACAGTGCT[C/T]GGGAGGGCTGGCTGGGATGGGAGTG
Phenotype
MIM: 138970 MIM: 607000
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSF3 PubMed Links

Gene Details

Gene
CSF3
Gene Name
colony stimulating factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000759.3 Intron NP_000750.1
NM_001178147.1 Intron NP_001171618.1
NM_172219.2 Intron NP_757373.1
NM_172220.2 Intron NP_757374.2
Gene
MED24
Gene Name
mediator complex subunit 24
There are no transcripts associated with this gene.

View Full Product Details