Product Details

SNP ID
rs2227460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:149374573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACCGTGCCTCCGGCAGGTCCAC[A/G]GGGATACGGCTGGGGTCGTGGTTGA
Phenotype
MIM: 604627
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL17B PubMed Links

Gene Details

Gene
IL17B
Gene Name
interleukin 17B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317987.1 712 Silent Mutation CCC,CCT P61P NP_001304916.1
NM_014443.2 712 Silent Mutation CCC,CCT P113P NP_055258.1
XM_011537622.2 712 Silent Mutation CCC,CCT P61P XP_011535924.1
XM_017009346.1 712 Silent Mutation CCC,CCT P178P XP_016864835.1
XM_017009347.1 712 Silent Mutation CCC,CCT P61P XP_016864836.1
XM_017009348.1 712 Silent Mutation CCC,CCT P61P XP_016864837.1
Gene
PCYOX1L
Gene Name
prenylcysteine oxidase 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301054.1 712 Intron NP_001287983.1
NM_001301057.1 712 Intron NP_001287986.1
NM_024028.3 712 Intron NP_076933.3
XM_011537680.1 712 Intron XP_011535982.1
XM_011537681.1 712 Intron XP_011535983.1
XM_011537682.2 712 Intron XP_011535984.1

View Full Product Details