Product Details

SNP ID
rs2972783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:42762318 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTTATGCAGTACATGACTGTATA[C/T]GTATTTTTTTATTAGACTATCAGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC152 PubMed Links

Gene Details

Gene
CCDC152
Gene Name
coiled-coil domain containing 152
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134848.1 Intron NP_001128320.1

View Full Product Details