Product Details

SNP ID
rs2270801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:132559022 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGGTCCCGCAGCTCCATTGTTGC[A/G]ATTGCTGAAAGATGAACTTCGTCAG
Phenotype
MIM: 614288
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACAD11 PubMed Links

Gene Details

Gene
ACAD11
Gene Name
acyl-CoA dehydrogenase family member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032169.4 2672 Silent Mutation ATC,ATT I764I NP_115545.3
Gene
NPHP3-ACAD11
Gene Name
NPHP3-ACAD11 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details