Product Details

SNP ID
rs2270818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157731431 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGGGCTATCTGGCCCTTTCCTTTC[A/C]GCGTGTAGAATGTGGGGCGCCTGTA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
THG1L PubMed Links

Gene Details

Gene
THG1L
Gene Name
tRNA-histidine guanylyltransferase 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317824.1 43 UTR 5 NP_001304753.1
NM_001317825.1 43 UTR 5 NP_001304754.1
NM_001317826.1 43 Intron NP_001304755.1
NM_017872.4 43 UTR 5 NP_060342.2
XM_017009603.1 43 Intron XP_016865092.1

View Full Product Details