Product Details
- SNP ID
-
rs2271278
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:40978309 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTACAGGAAAGCCTGGGGAAAAAT[C/T]GATTTTCAACCAAGATTAACAAGGG
- Phenotype
-
MIM: 616679
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT40
PubMed Links
Gene Details
- Gene
- KRT40
- Gene Name
- keratin 40
View Full Product Details