Product Details
- SNP ID
-
rs2272846
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:50201536 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACCTGGGCGAGGTGTGCACGGCGA[A/C]CGGCGAGCGCTGGGAGCTGCAGCTC
- Phenotype
-
MIM: 607917
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SELO
PubMed Links
Gene Details
- Gene
- SELO
- Gene Name
- selenoprotein O
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_031454.1 |
558 |
Missense Mutation |
AAC,ACC |
N167T |
NP_113642.1 |
- Gene
- TRABD
- Gene Name
- TraB domain containing
There are no transcripts associated with this gene.
View Full Product Details