Product Details

SNP ID
rs2278652
Assay Type
Validated
NCBI dbSNP Submissions
33
Location
Chr.1:211577683 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
CAAAGTTGCTGCAGCCTGTAAAGAC[A/G]GCCAAGTGCGGGTGTGGACTGCAGC
Phenotype
MIM: 609521
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC30A1 PubMed Links

Gene Details

Gene
SLC30A1
Gene Name
solute carrier family 30 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021194.2 Intron NP_067017.2

View Full Product Details