Product Details

SNP ID
rs2278999
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50466719 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAACCGAAGAGGGAGGCAGCCGTA[T/C]TGGACAAACTCCTGCCAGGCAGCTG
Phenotype
MIM: 614545 MIM: 160793
Polymorphism
T/C, Transition Substitution
Allele Nomenclature
Literature Links
EMC10 PubMed Links
Additional Information
For this assay, SNP(s) [rs2308271] are located under a probe and SNP(s) [rs6509476] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
EMC10
Gene Name
ER membrane protein complex subunit 10
There are no transcripts associated with this gene.

Gene
FAM71E1
Gene Name
family with sequence similarity 71 member E1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308429.1 944 Intron NP_001295358.1
NM_138411.2 944 Intron NP_612420.1
XM_011526405.2 944 Missense Mutation AAT,AGT N326S XP_011524707.2
XM_011526406.2 944 Missense Mutation AAT,AGT N310S XP_011524708.2
Gene
MYBPC2
Gene Name
myosin binding protein C, fast type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004533.3 944 Intron NP_004524.3

View Full Product Details