Product Details

SNP ID
rs2280345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65381839 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTACCCAGAGCCCAAATCATATATA[A/G]TGCTAGTGAGAAATGGCCACTTTCT
Phenotype
MIM: 604184 MIM: 616810
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGDCC3 PubMed Links

Gene Details

Gene
IGDCC3
Gene Name
immunoglobulin superfamily DCC subclass member 3
There are no transcripts associated with this gene.

Gene
IGDCC4
Gene Name
immunoglobulin superfamily DCC subclass member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020962.2 5940 UTR 3 NP_066013.1
XM_011521845.2 5940 UTR 3 XP_011520147.1
XM_011521846.2 5940 UTR 3 XP_011520148.1
XM_011521847.1 5940 UTR 3 XP_011520149.1
XM_017022448.1 5940 UTR 3 XP_016877937.1
XM_017022449.1 5940 Intron XP_016877938.1

View Full Product Details