Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001242314.1 | 5055 | UTR 3 | NP_001229243.1 | ||
NM_001242317.1 | 5055 | UTR 3 | NP_001229246.1 | ||
NM_004721.4 | 5055 | UTR 3 | NP_004712.1 | ||
XM_011513310.2 | 5055 | Intron | XP_011511612.1 | ||
XM_017007456.1 | 5055 | UTR 3 | XP_016862945.1 | ||
XM_017007457.1 | 5055 | Intron | XP_016862946.1 | ||
XM_017007458.1 | 5055 | Intron | XP_016862947.1 | ||
XM_017007459.1 | 5055 | Intron | XP_016862948.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_080652.3 | 5055 | Intron | NP_542383.1 | ||
XM_017007437.1 | 5055 | Intron | XP_016862926.1 |