Product Details

SNP ID
rs2280677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126394284 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTTCAAGCAGCTGGTGGATCTGC[A/G]CGCTCAAAACTTGCCCTGGTCTGTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC37-AS1 PubMed Links

Gene Details

Gene
CCDC37-AS1
Gene Name
CCDC37 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
CFAP100
Gene Name
cilia and flagella associated protein 100
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182628.2 Intron NP_872434.2
XM_006713623.2 Intron XP_006713686.1
XM_017006320.1 Intron XP_016861809.1
XM_017006321.1 Intron XP_016861810.1
XM_017006322.1 Intron XP_016861811.1
XM_017006323.1 Intron XP_016861812.1
XM_017006324.1 Intron XP_016861813.1
XM_017006325.1 Intron XP_016861814.1
XM_017006326.1 Intron XP_016861815.1
XM_017006327.1 Intron XP_016861816.1

View Full Product Details