Product Details

SNP ID
rs3001342
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:2512304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGAGTGGGACCCAGGGAGGGCTGA[G/A]AACCCAGGCATTTCCCTCCAGCCCC
Phenotype
MIM: 606162 MIM: 612836
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
PANK4 PubMed Links

Gene Details

Gene
PANK4
Gene Name
pantothenate kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018216.2 Intron NP_060686.2
XM_011541708.2 Intron XP_011540010.2
XM_017001633.1 Intron XP_016857122.1
Gene
PLCH2
Gene Name
phospholipase C eta 2
There are no transcripts associated with this gene.

View Full Product Details