Product Details

SNP ID
rs3020937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153517722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGCACTCGGCCGCAGCCCTCGC[C/T]CGCCGCAGAGGGTCGCGTCCCGCGC
Phenotype
MIM: 300014 MIM: 301870
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP2B3 PubMed Links

Gene Details

Gene
ATP2B3
Gene Name
ATPase plasma membrane Ca2+ transporting 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001344.2 45 Intron NP_001001344.1
NM_021949.3 45 Intron NP_068768.2
XM_005274689.3 45 UTR 5 XP_005274746.1
XM_005274690.3 45 UTR 5 XP_005274747.1
XM_005274691.3 45 UTR 5 XP_005274748.1
XM_005274692.3 45 UTR 5 XP_005274749.1
XM_011531174.2 45 UTR 5 XP_011529476.1
XM_011531177.2 45 UTR 5 XP_011529479.1
XM_017029553.1 45 Intron XP_016885042.1
Gene
BGN
Gene Name
biglycan
There are no transcripts associated with this gene.

View Full Product Details