Product Details
- SNP ID
-
rs2352823
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:240436110 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGAGTGAGGGCGCGGCGCCGGGAA[C/T]CCGGGCCTGGCCGGGCTTTGGGCTC
- Phenotype
-
MIM: 600395
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GPC1
PubMed Links
Gene Details
- Gene
- GPC1
- Gene Name
- glypican 1
View Full Product Details