Product Details

SNP ID
rs2355870
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:231367134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTACTGATTTTGGCATTTGAATTC[A/G]TAATTCTTTGTATTAATTTTCTTCA
Phenotype
MIM: 606425
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EGLN1 PubMed Links

Gene Details

Gene
EGLN1
Gene Name
egl-9 family hypoxia inducible factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022051.2 Intron NP_071334.1
XM_005273166.4 Intron XP_005273223.1
XM_005273167.4 Intron XP_005273224.1

View Full Product Details