Product Details

SNP ID
rs2364600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53269289 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGCTGCACAAGGAAGCCCAGAGG[C/T]CACACTCCGCCTTGCTCAGCCCCTC
Phenotype
MIM: 604143
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESPL1 PubMed Links

Gene Details

Gene
ESPL1
Gene Name
extra spindle pole bodies like 1, separase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012291.4 438 Missense Mutation GCC,GTC A116V NP_036423.4
XM_006719705.3 438 Missense Mutation GCC,GTC A116V XP_006719768.1
XM_011539024.2 438 Missense Mutation GCC,GTC A116V XP_011537326.1
XM_011539025.2 438 Missense Mutation GCC,GTC A116V XP_011537327.1
XM_017020253.1 438 UTR 5 XP_016875742.1

View Full Product Details