Product Details

SNP ID
rs3087876
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:10942767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAGGAAACCCGGGCTACCGCTGC[A/G]CCTTGCTGCATGCCTCTTTGGGACT
Phenotype
MIM: 600005 MIM: 611303
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CIITA PubMed Links

Gene Details

Gene
CIITA
Gene Name
class II, major histocompatibility complex, transactivator
There are no transcripts associated with this gene.

Gene
CLEC16A
Gene Name
C-type lectin domain family 16 member A
There are no transcripts associated with this gene.

Gene
DEXI
Gene Name
Dexi homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014015.3 241 Intron NP_054734.2
XM_011522455.2 241 Intron XP_011520757.1
XM_017023171.1 241 UTR 5 XP_016878660.1

View Full Product Details