Product Details

SNP ID
rs2433654
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:184599215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGTGGGGTGAGAATTGGGTGTA[A/G]AAAAAGGAAGGGGTCTTAGGGAAGC
Phenotype
MIM: 612282
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105373780 PubMed Links

Gene Details

Gene
LOC105373780
Gene Name
proline-rich protein 18-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011512285.1 Intron XP_011510587.1
Gene
ZNF804A
Gene Name
zinc finger protein 804A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194250.1 Intron NP_919226.1

View Full Product Details