Product Details

SNP ID
rs2508665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:117328073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACCCGAGGCACGCTCTCGAGCC[A/T]ACGAGCGTGATGCGCTCGAGTGTGG
Phenotype
MIM: 614848
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CEP164 PubMed Links

Gene Details

Gene
CEP164
Gene Name
centrosomal protein 164
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271933.1 140 Intron NP_001258862.1
NM_014956.4 140 Intron NP_055771.4
XM_005271453.1 140 UTR 5 XP_005271510.1
XM_005271456.1 140 UTR 5 XP_005271513.1
XM_005271457.1 140 UTR 5 XP_005271514.1
XM_006718788.1 140 UTR 5 XP_006718851.1
XM_006718794.1 140 UTR 5 XP_006718857.1
XM_011542674.1 140 UTR 5 XP_011540976.1
XM_011542690.1 140 UTR 5 XP_011540992.1
XM_017017364.1 140 Intron XP_016872853.1
XM_017017365.1 140 Intron XP_016872854.1
XM_017017366.1 140 Intron XP_016872855.1
XM_017017367.1 140 UTR 5 XP_016872856.1
XM_017017368.1 140 UTR 5 XP_016872857.1
XM_017017369.1 140 UTR 5 XP_016872858.1
XM_017017370.1 140 UTR 5 XP_016872859.1
XM_017017371.1 140 UTR 5 XP_016872860.1
XM_017017372.1 140 UTR 5 XP_016872861.1
XM_017017373.1 140 UTR 5 XP_016872862.1
XM_017017374.1 140 Intron XP_016872863.1
XM_017017375.1 140 UTR 5 XP_016872864.1
XM_017017376.1 140 UTR 5 XP_016872865.1
XM_017017377.1 140 UTR 5 XP_016872866.1
XM_017017378.1 140 UTR 5 XP_016872867.1
XM_017017379.1 140 UTR 5 XP_016872868.1
XM_017017380.1 140 Intron XP_016872869.1
XM_017017381.1 140 UTR 5 XP_016872870.1
XM_017017382.1 140 Intron XP_016872871.1
XM_017017383.1 140 UTR 5 XP_016872872.1
XM_017017384.1 140 Intron XP_016872873.1
XM_017017385.1 140 Intron XP_016872874.1
XM_017017386.1 140 UTR 5 XP_016872875.1

View Full Product Details