Product Details

SNP ID
rs2513737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62352023 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTTAGGCCTGCTGGCAACAAATT[C/G]TTTCCATTTGCCCTTATCTGTGTTA
Phenotype
MIM: 609212
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ASRGL1 PubMed Links

Gene Details

Gene
ASRGL1
Gene Name
asparaginase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083926.1 Intron NP_001077395.1
NM_025080.3 Intron NP_079356.3
XM_005274305.3 Intron XP_005274362.1
XM_005274306.2 Intron XP_005274363.1
XM_011545265.2 Intron XP_011543567.1
XM_011545266.2 Intron XP_011543568.1
XM_017018354.1 Intron XP_016873843.1

View Full Product Details