Product Details

SNP ID
rs2484532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:131127876 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTAATGGAAGCCTTTCTAGGTTTT[C/T]TCCTTGGTCCTTTGTATTTCTCCTT
Phenotype
MIM: 114350
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AIF1L PubMed Links

Gene Details

Gene
AIF1L
Gene Name
allograft inflammatory factor 1 like
There are no transcripts associated with this gene.

Gene
NUP214
Gene Name
nucleoporin 214
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318324.1 Intron NP_001305253.1
NM_001318325.1 Intron NP_001305254.1
NM_005085.3 Intron NP_005076.3
XM_005272216.3 Intron XP_005272273.1
XM_005272218.3 Intron XP_005272275.1
XM_005272219.3 Intron XP_005272276.1
XM_006717292.3 Intron XP_006717355.1
XM_011519053.2 Intron XP_011517355.1
XM_011519055.2 Intron XP_011517357.1
XM_017015167.1 Intron XP_016870656.1

View Full Product Details