Product Details

SNP ID
rs2485447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33290603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGCGCCTCCTGTCTCAGGTATT[G/T]TCCCGGCCCGAGCGGGACTGGGCCC
Phenotype
MIM: 610900 MIM: 603255
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CHMP5 PubMed Links

Gene Details

Gene
CHMP5
Gene Name
charged multivesicular body protein 5
There are no transcripts associated with this gene.

Gene
NFX1
Gene Name
nuclear transcription factor, X-box binding 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318758.1 Intron NP_001305687.1
NM_002504.5 Intron NP_002495.2
NM_147134.3 Intron NP_667345.1

View Full Product Details