Product Details

SNP ID
hCV16039612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:20886825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAATCAACAATCCTATTTTAAC[T/C]CAATTAAAAAATAGAAAATCTTGAT
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CCDC144NL PubMed Links

Gene Details

Gene
CCDC144NL
Gene Name
coiled-coil domain containing 144 family, N-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004306.2 Intron NP_001004306.1
Gene
CCDC144NL-AS1
Gene Name
CCDC144NL antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details