Product Details

SNP ID
rs2568492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78446778 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTACCAGGGGATTGGCTTTTGCGTC[A/G]CTGCTGTAGCCAAGGAGGCCACAGG
Phenotype
MIM: 147582
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IREB2 PubMed Links

Gene Details

Gene
IREB2
Gene Name
iron responsive element binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320941.1 Intron NP_001307870.1
NM_001320942.1 Intron NP_001307871.1
NM_001320943.1 Intron NP_001307872.1
NM_004136.3 Intron NP_004127.1

View Full Product Details