Product Details

SNP ID
rs2633462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:127886908 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCAGCTGCTTCCTAGCTGCCAATA[A/C]CCTTCATTCAGCTCAAGAAAAACAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FLJ37505 PubMed Links

Gene Details

Gene
FLJ37505
Gene Name
uncharacterized LOC400087
There are no transcripts associated with this gene.

Gene
LINC00508
Gene Name
long intergenic non-protein coding RNA 508
There are no transcripts associated with this gene.

View Full Product Details