Product Details

SNP ID
rs2572926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142786442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGCCACAGCCGCCCACCCGCCC[A/G]TCCCCTTGGCCCAGCCCCTCACCTG
Phenotype
MIM: 606204 MIM: 606110
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LY6D PubMed Links

Gene Details

Gene
LY6D
Gene Name
lymphocyte antigen 6 complex, locus D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003695.2 Intron NP_003686.1
Gene
LYNX1
Gene Name
Ly6/neurotoxin 1
There are no transcripts associated with this gene.

View Full Product Details