Product Details

SNP ID
rs2668712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:46264985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAGAGCACGTGAAAGAAGGGGA[C/T]CTGAGGACATCTATGAACAGGGCCC
Phenotype
MIM: 616555
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL17B PubMed Links

Gene Details

Gene
ARL17B
Gene Name
ADP ribosylation factor like GTPase 17B
There are no transcripts associated with this gene.

Gene
LRRC37A
Gene Name
leucine rich repeat containing 37A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014834.4 Intron NP_055649.4
XM_005257879.4 Intron XP_005257936.1
XM_005257880.4 Intron XP_005257937.1
XM_005257882.4 Intron XP_005257939.1
XM_005257883.4 Intron XP_005257940.1
XM_005257884.4 Intron XP_005257941.1
XM_006722210.3 Intron XP_006722273.1
XM_006722211.3 Intron XP_006722274.1
XM_011525539.2 Intron XP_011523841.1
XM_017025472.1 Intron XP_016880961.1
XM_017025473.1 Intron XP_016880962.1

View Full Product Details