Product Details

SNP ID
rs2747414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29658216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTAGGTTTTTATAAAGGAACCCG[G/T]CTGTCACGTGCAGAGGAGAAATAAA
Phenotype
MIM: 159465
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MOG PubMed Links

Gene Details

Gene
MOG
Gene Name
myelin oligodendrocyte glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008228.2 Intron NP_001008229.1
NM_001008229.2 Intron NP_001008230.1
NM_001170418.1 Intron NP_001163889.1
NM_002433.4 Intron NP_002424.3
NM_206809.3 Intron NP_996532.2
NM_206810.3 Intron NP_996533.2
NM_206811.3 Intron NP_996534.2
NM_206812.3 Intron NP_996535.2
NM_206814.5 Intron NP_996537.3
XM_005249131.3 Intron XP_005249188.1

View Full Product Details