Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001318807.1 | 2018 | UTR 3 | NP_001305736.1 | ||
NM_014430.2 | 2018 | UTR 3 | NP_055245.2 | ||
XM_011536659.2 | 2018 | UTR 3 | XP_011534961.1 | ||
XM_017021221.1 | 2018 | UTR 3 | XP_016876710.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001286367.1 | 2018 | UTR 3 | NP_001273296.1 | ||
NM_174913.2 | 2018 | UTR 3 | NP_777573.1 | ||
XM_005267385.1 | 2018 | UTR 3 | XP_005267442.1 | ||
XM_011536526.1 | 2018 | UTR 3 | XP_011534828.1 | ||
XM_011536527.2 | 2018 | Intron | XP_011534829.1 |