Product Details

SNP ID
rs2835846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:37628063 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAAAAACTCTGAAAAGGAGGGAG[A/C]ATCTGATTTTCAGGGTTACTACATT
Phenotype
MIM: 600877
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KCNJ6 PubMed Links

Gene Details

Gene
KCNJ6
Gene Name
potassium voltage-gated channel subfamily J member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002240.4 Intron NP_002231.1

View Full Product Details