Product Details

SNP ID
rs2131975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40090927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTGTCGGTGGGGTCTTTTGAAA[A/G]AAGAGCTGACAAAGGCACAGGCTGC
Phenotype
MIM: 606266
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BMF PubMed Links

Gene Details

Gene
BMF
Gene Name
Bcl2 modifying factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003940.1 1499 UTR 3 NP_001003940.1
NM_001003942.1 1499 UTR 3 NP_001003942.1
NM_001003943.2 1499 UTR 3 NP_001003943.1
NM_033503.3 1499 UTR 3 NP_277038.1

View Full Product Details