Product Details
- SNP ID
-
rs2893376
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
29
- Location
-
Chr.1:97991747 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTAAATGTTTGACAAGTTTGACAA[C/G]CTAAAACTAGACAACGAAATTGCTG
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR137HG
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141221548] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR137HG
- Gene Name
- MIR137 host gene
There are no transcripts associated with this gene.
View Full Product Details