Product Details
- SNP ID
-
rs2169925
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:43621495 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGTTGGCAGGGAGCAAGTGGCAG[A/T]CAGCACCCCTCTCTCCCTAGCCTAT
- Phenotype
-
MIM: 607434
MIM: 603968
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GTPBP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113592918] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GTPBP2
- Gene Name
- GTP binding protein 2
- Gene
- MAD2L1BP
- Gene Name
- MAD2L1 binding protein
There are no transcripts associated with this gene.
- Gene
- POLH
- Gene Name
- DNA polymerase eta
View Full Product Details