Product Details

SNP ID
rs2794813
Assay Type
Functionally tested
NCBI dbSNP Submissions
45
Location
Chr.1:237882290 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
AAGTCTGAAATTTTTGTATCAGTTT[C/T]TATTCTATTCTCAGTTGAGTTATAC
Phenotype
MIM: 613514
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100130331 PubMed Links

Gene Details

Gene
LOC100130331
Gene Name
POTE ankyrin domain family, member F pseudogene
There are no transcripts associated with this gene.

Gene
ZP4
Gene Name
zona pellucida glycoprotein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021186.4 Intron NP_067009.1

View Full Product Details