Product Details

SNP ID
rs2253815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:1565561 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACATCCACATTCATTCATCTGCTC[A/G]TCATTTAAGAAATGTTTCCTGAGAG
Phenotype
MIM: 603889
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIRPB1 PubMed Links

Gene Details

Gene
SIRPB1
Gene Name
signal regulatory protein beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005260641.3 Intron XP_005260698.1
XM_005260643.3 Intron XP_005260700.1
XM_005260644.3 Intron XP_005260701.1
XM_011529130.2 Intron XP_011527432.1
XM_011529134.2 Intron XP_011527436.1
XM_017027577.1 Intron XP_016883066.1
XM_017027578.1 Intron XP_016883067.1
XM_017027579.1 Intron XP_016883068.1
XM_017027580.1 Intron XP_016883069.1
XM_017027581.1 Intron XP_016883070.1
XM_017027582.1 Intron XP_016883071.1
XM_017027583.1 Intron XP_016883072.1
XM_017027584.1 Intron XP_016883073.1
XM_017027585.1 Intron XP_016883074.1
Gene
SIRPD
Gene Name
signal regulatory protein delta
There are no transcripts associated with this gene.

View Full Product Details