Product Details

SNP ID
rs2859311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1620690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGTCTATTAGCCGCGTGCCTTT[C/T]GGTGAGTCATTTAACTTCTCACCTT
Phenotype
MIM: 614620
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRAMP1 PubMed Links

Gene Details

Gene
CRAMP1
Gene Name
cramped chromatin regulator homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020825.3 Intron NP_065876.3
Gene
IFT140
Gene Name
intraflagellar transport 140
There are no transcripts associated with this gene.

View Full Product Details