Product Details
- SNP ID
-
rs2838318
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:43571944 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGGAGAGAGAGGGCTGAATCAGAGC[A/G]ACTAGGTCCAAAGCCGAGGGAACCA
- Phenotype
-
MIM: 604554
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
H2BFS
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77836310] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- H2BFS
- Gene Name
- H2B histone family member S (pseudogene)
There are no transcripts associated with this gene.
- Gene
- HSF2BP
- Gene Name
- heat shock transcription factor 2 binding protein
View Full Product Details